Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Scotch™ Super 33+ Vinyl Electrical Tape
3M™ Scotch™ Super 33+ Vinyl Electrical Tape features a combination of pressure-sensitive, rubber-resin adhesive and PVC backing that offers electrical and mechanical protection
Supplier: 3M™ 7010349613
Description
- 7 mil thick
- Resists UV rays, abrasion, moisture, alkalis, solvents and acids
- Flame-retardant
- Inhibits the corrosion of electrical conductors
- Provides electrical insulation for all wires and cables rated up to 600V
- Made of durable PVC
- Compatible with solid dielectric cable insulation, synthetic cable insulations, jackets and splicing compounds
Specifications
UL Listed, CSA Certified and RoHS 2011/65/EU Compliant | |
Rubber | |
Automotive, Communications, Design and Construction, Electronics, Energy, Manufacturing, Mining, Oil and Gas, Transportation | |
100 Rolls/Cs. | |
18°C to 105°C |
Black | |
Vinyl Electrical Tape | |
33+-3/4X66FT-1.5 | |
0°F to 221°F | |
66 ft. x 3/4 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction