Learn More
World Precision Instrument RTV Sealant
Supplier: World Precision Instrument 7128
Because it is non-corrosive, this material is ideal for use on metal, for encapsulating small circuits on connectors. After cure, will withstand -55° to 200 °C. No mixing required.