Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rat Progesterone ELISA Kit (Colorimetric)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Assay has high sensitivity and specificity for detection of rat PROG
Supplier: Novus Biologicals™ NBP2601271KIT
Description
The Rat Progesterone ELISA Kit (Colorimetric) measures rat Progesterone in serum, plasma, cell culture supernates, tissue homogenates.Specifications
1ng/mL to 30ng/mL | |
ELISA Kit (Colorimetric) | |
For detection of rat PROG | |
ELISA | |
1 Kit | |
50 to 100 μL | |
Pregn 4 ene 3 20 dione |
0.2ng/mL | |
Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates | |
Rat | |
Instruction manual, Stop Solution, Standard, Substrate B, Substrate A, Wash Buffer (20 x concentrate), HRP-conjugate, Adhesive Strip (For 96 wells), Antibody, Assay plate | |
Signal Transduction | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
Progesterone |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction