Learn More
Fisherbrand™ PTFE Dippers
SDPSCI_ED
Totally inert, reusable.
 View more versions of this product
Catalog No. S37115
Explore available promotions
Add to Cart
Description
Description
  • Extendable screw-in shafts have a steel core for rigidity
  • Fully encapsulated in PTFE
Specifications
Specifications
Dipper
500 mL
3.14 in.
29 in.
4.13 in.
10.5 cm
3.19 in.
PTFE
4.13 x 3.19 x 29 in. (10.5 x 8.1 x 73.6 cm)
8 cm
73.6 cm
24 in.
61 cm
8.1 cm
Product Suggestions
Product Suggestions