Learn More
Prolume Ltd CAT#3692 Prolume Purple- SOL (in vitro) 1 tube
Supplier: Prolume Ltd 36921
Methoxy e-Coelenterazine (Prolume Purple) formulated to be water-soluble for in vitro applications with Renilla Luciferase.Prolume Purple exhibits an up to 13-fold higher luminescence than CTZ400a (“deep blue C”).