Learn More
Thermo Scientific Post-Column Cooler | Stainless Steel | 1 EA
Supplier: Thermo Scientific 6730.0008
Minimize noise and drift in high temperature chromatography applications. Efficient precolumn heating and postcolumn cooling technology. Reduced system backpressures and typically sharper peaks.