Learn More
Biolegend PE/FireTM 810 anti-human CD2
Supplier: Biolegend 302860
PE/Fire™ 810 anti-human CD27 [O323]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests