Learn More
MilliporeSigma™ Opticap™ Sterile XL600 Capsule Filters
ideal for sterile filtration of cell culture media, protein solutions and other high fouling fluids
Supplier:  MilliporeSigma™ KHVES006HH3
 View more versions of this product
Catalog No. KHVES006HH3
Add to Cart
Description
Description
Specifications
Specifications
Opticap Sterile XL 600 Millipore Express SHR with Prefilter
Sterile
Inlet/Outlet Connection: 0.56 in. (14mm) Hose Barb
16.5 cm
0.5/0.2 μm
0.59 m2
6.5 in.
Product Suggestions
Product Suggestions