Learn More
MilliporeSigma™ Opticap™ Sterile XL600 Capsule Filters
ideal for sterile filtration of cell culture media, protein solutions and other high fouling fluids
Supplier:  MilliporeSigma™ KHVES006FF3
 View more versions of this product
Catalog No. KHVES006FF3
Add to Cart
Description
Description
Specifications
Specifications
Opticap Sterile XL 600 Millipore Express SHR with Prefilter
Sterile
Inlet/Outlet Connection: 0.75 in. (19mm) Sanitary Flange
16.5 cm
0.5/0.1 μm
0.59 m2
6.5 in.
Product Suggestions
Product Suggestions