Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse TNF-alpha DuoSet ELISA
Mouse TNF-alpha DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$504.60 - $916.40
Specifications
Product Type | ELISA Development Kit |
---|---|
Quantity | 1 Kit |
Species | Mouse |
Conjugate | Biotin |
Assay Range | 31.20 to 2000 pg/mL |
Description
The Mouse TNF-alpha DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse TNF-alpha.Specifications
ELISA Development Kit | |
Mouse | |
31.20 to 2000 pg/mL | |
ELISA | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
21926 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title