Learn More
Sino Biological MAP1D / Methionine Aminopeptidase 1D Antibody, Mouse MAb 100uL
Supplier: Sino Biological 10883MM04100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MAP1D (rh MAP1D; Catalog#10883-H08B; NP_954697.1; Arg 44-Ala 335). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
This item is not returnable.
View return policy