Learn More
Med Vet International LIQUIVET 2 SEC VET USE 3G EACH
Supplier: Med Vet International VVG3
LiquiVet monomeric formulation that forms a thin, waterproof adhesion to bond tissue in a variety of vet procedures. Fast-acting Cyanoacrylate based tissue adhesive for wound care. Polymerizes in 2 seconds (VG3). Ideal for skin closure. Excellent for quick repair of minor lacerations, abrasions, and other minor wounds. Superior performance in the time necessary to close a wound. For external lacerations, not to be used internally. May be alternative to products like ZipStitch instant stitches and Strips For skin closure Best suited for small, superficial lacerations Quick and virtually painless to use Wears off in the days and weeks Reduce the chances of an infection
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.