Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Tartaric Acid (Powder/Certified ACS), Fisher Chemical

Catalog Number A315500
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
87-69-4
C4H6O6
150.09
MFCD00064207
FEWJPZIEWOKRBE-UHFFFAOYNA-N
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
444305
CHEBI:15671
2,3-dihydroxybutanedioic acid
OC(C(O)C(O)=O)C(O)=O
87-69-4
C4H6O6
150.09
MFCD00064207
FEWJPZIEWOKRBE-UHFFFAOYNA-N
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
444305
CHEBI:15671
2,3-dihydroxybutanedioic acid
OC(C(O)C(O)=O)C(O)=O
pH 1.6
Melting Point 168°C
Color White
Assay Percent Range ≥99.0 %
Linear Formula HOOC(CHOH)2COOH
Formula Weight 150.09
Percent Purity ≥99.0%
Grade Certified ACS
Quantity 500 g
Ignition Residue 0.02% max.
Packaging Poly Bottle
Physical Form Solid
Vapor Pressure <0.1 mbar at 20°C
Chemical Name or Material L-(+)-Tartaric Acid
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit