Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Autoclavable and used for multiple injections
$766.50 - $925.50
Catalog Number | Mfr. No. | Color | Pre-slit | dimClosureSize | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Pre-slit | dimClosureSize | Price | Quantity & Availability | |||
03-341-605
|
DWK Life Sciences
73813BK8425 |
Black | Yes |
Case of 1000 for $925.50
|
|
||||
03-341-606
|
DWK Life Sciences
73812WH8425 |
White | No |
Case of 1000 for $766.50
|
|
||||
03-341-604
|
DWK Life Sciences
73812BK8425 |
Black | No |
Case of 1000 for $766.50
|
|
||||
03-341-608
|
DWK Life Sciences
73812BK9425 |
Black | No |
Case of 1000 for $766.50
|
|
||||
03-341-609
|
DWK Life Sciences
73813BK9425 |
Black | Yes |
Case of 1000 for $925.50
|
|
||||
03-341-613
|
DWK Life Sciences
73813BK13425 |
Black | Yes |
Case of 1000 for $925.50
|
|
||||
03-341-611
|
DWK Life Sciences
73813BL9425 |
Blue | Yes |
Case of 1000 for $925.50
|
|
||||
Description
- Manufactured using the highest quality raw materials
- Designed for an ideal fit with Kimble Chase autosampler vials
- Convenient assembly of caps and liners
- Available with standard or pre-slit septa
Specifications
Polypropylene | |
Autoclavable |