Learn More
Kimble™ KIMAX™ Heavy-duty Filtering Flask
The plastic sidearm conveniently detaches from the flask
Supplier: Kimble™ 27065250/EMD
Description
- Made of heavy-duty Kimax borosilicate glass
- Has a detachable and autoclavable plastic sidearm
- Sidearm attached by neoprene grommet
Order Info
Sold in packs of 2
Specifications
250 mL | |
Borosilicate Glass | |
75 to 250 mL |