Learn More
MilliporeSigma™ Immobilon Crescendo Western HRP substrate
For use on immunoblots
Supplier:  MilliporeSigma™ WBLUR0020
Catalog No. WBLUR0020
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Western blotting, dot/slot blotting
Specifications
Specifications
Chemiluminescent
20 mL
2°C to 8°C, Stable for 1 year at 4°C from date of shipment. Contains hydrogen peroxide. Use in well ventilated area.
Immunoblot (Western)
HRP