Learn More
Foxx Life Sciences EZWaste® HPLC Universal Stackable VersaCap® 70mm(S70) Solvent Waste System, Cap-Only W/Exhaust Filter, Six(6x) Ports OD Tubing -3.2 mm (1/8"") or 1.6 mm(1/16""), Three(3x) Hose Barb 3.2mm (1/8"") or 9.5mm(3/8""), Three(3x) OD Tubing 12.7 mm (1/2""), 1/EA
Supplier: Foxx Life Sciences 3300B60FLS
Foxx Life Sciences EZwaste® Universal Stackable HPLC VersaCap® GL45 Solvent Waste System is designed to be an affordable solution for HPLC liquid waste collection & disposal. System features an activated carbon filter which traps volatile organic compounds from being released into the air. The EZwaste® HPLC Solvent Waste system also features a closed loop design which stops leaks & improves overall lab safety. In EZwaste® Universal Stackable design you can add multiple manifolds for increased ports, conveniently as per requirement.Each EZwaste® Assy. includes a VersaCap® GL45, Universal Stackable Manifold w/ 6-3-3 Ports, 6x fittings of 3.2mm (1/8") OD Tubing, 6x fittings of 1.6 mm (1/16") OD Tubing, 3x fittings of 3.2mm (1/8") Hose Barb, 3x fittings of 9.5mm (3/8") Hose Barb, 3x fittings of 12.7mm (1/2") OD Tubing , 1 carbon Exhaust Filter & all suitable plugs/adapters for blocking of unused ports. * Carbon exhaust filter life may very.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.