Learn More
Andwin Scientific Battery powered densitometer, 110V, 75 x 115 x 165mm
SDP
Supplier:  Andwin Scientific DEN1B
Encompass_Preferred
Grant Instruments. The Grant bio DEN-1 and DEN-1B are a compact benchtop densitometers (suspension turbidity meter) for measuring turbidity of cell suspensions in a variety of life science applications. The densitometers are designed and factory calibrated to measure turbidity in the range of 0.5 to 4.0 McFarland units (DEN-1) 0.0 to >7.5 McFarland units (DEN-1B) with a small standard deviation. If required, it can deliver a wider measurement range (up to 15.00 McFarland units), but with a greater standard deviation. Measurement range: 0.5 to 4.0 McFarland units (DEN-1), 0.0 to >7.5 McFarland units (DEN-1B)Measurement time : 1 second. Precision: ±3%.
Catalog No. NC1154909