Learn More
Thermo Scientific Chemicals Glutamic pyruvic transaminase, porcine heart
Description
It is used for research and clinical diagnostic assays for liver disease. Alanine transaminase (GPT/ALT) is used in multi-analyte clinical chemistry controls and calibrators for major IVD manufacturers. The alanine aminotransferase (ALT) blood test is typically used to detect liver injury. Some of the other applications include Diagnostic Controls, calibrators & standards, immunoassays, clinical chemistry, testing/assay validation, life Science, manufacturing, validation Studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Packaging | Plastic bottle |
Quantity | 1 kU |
Beilstein | 3562140 |
Solubility Information | Dissolves readily at 5mg/ml in 0.05M potassium phosphate pH 7.5 to give a clear,pale-green solution. Soluble in distilled water or dilute buffer. |
Physical Form | Lyophilized Powder |
Chemical Name or Material | Glutamic pyruvic transaminase |
Safety and Handling
EINECSNumber | 232-561-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.