Learn More
Bioassay Systems EnzyFluo™ Glucose Uptake Assay Kit. For quantitative determination of glucose uptake in whole cells and evaluation of effects of ligands or drugs on glucose transport.
SDP
Supplier:  Bioassay Systems EFGU100
Encompass_Preferred
For quantitative determination of glucose uptake in whole cells and evaluation of effects of ligands or drugs on glucose transport. Key Features: Safe. No radioactive material is used. Sensitive and Accurate. Detection limit of 0.1 µM and linearity up to 5 µM 2-DG6P. Simple and Convenient. Can be automated as a medium throughput assay for glucose transport in cells. Method: FL530/585nm. Samples: Cell culture. Species: Mammalian. Procedure: Assay takes Approximately 2 hrs. Kit size: 100 tests. Detection limit: 0.1 µM.
Catalog No. 50-225-5878