Learn More
United Scientific™ Friction Rods and Pads
Ideal tools for demonstrating static electricity.
Supplier: United Scientific™ FRHR06
Description
- Friction rods and pads made from various materials.
- All the essential materials for student exploration in electrostatics
- Topics include static electricity, polarity, and friction studies
- Use a variety of friction rods, friction pads, and pith balls
- The combination of materials will determine the charge distribution after contact is made
Specifications
Friction Rods and Pads | |
Rubber | |
6 in. |
Hard Rod | |
Friction Demonstrations |