Learn More
Ampco Safety Tools Double Open Wrench (Metric)
AMPCO™ Double Open Wrench performs in high-risk and demanding environments.
$80.60 - $1718.00
Specifications
Product Type | Double Open Wrench |
---|---|
Material | Aluminum Bronze |
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog Number | Mfr. No. | Drive Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Drive Size | Price | Quantity & Availability | |||||
19-174-782
|
Ampco Safety Tools
WO10X11 |
10 x 11 mm |
Each for $145.90
|
|
|||||
19-174-783
|
Ampco Safety Tools
WO10X12 |
10 x 12 mm |
Each for $141.80
|
|
|||||
19-174-784
|
Ampco Safety Tools
WO10X13 |
10 x 13 mm |
Each for $141.80
|
|
|||||
19-174-785
|
Ampco Safety Tools
WO10X14 |
10 x 14 mm |
Each for $141.80
|
|
|||||
19-174-797
|
Ampco Safety Tools
WO11X12 |
11 x 12 mm |
Each for $141.80
|
|
|||||
19-174-798
|
Ampco Safety Tools
WO11X13 |
11 x 13 mm |
Each for $154.10
|
|
|||||
19-174-799
|
Ampco Safety Tools
WO11X14 |
11 x 14 mm |
Each for $154.10
|
|
|||||
19-174-800
|
Ampco Safety Tools
WO12X13 |
12 x 13 mm |
Each for $154.10
|
|
|||||
19-174-801
|
Ampco Safety Tools
WO12X14 |
12 x 14 mm |
Each for $157.10
|
|
|||||
19-174-804
|
Ampco Safety Tools
WO13X14 |
13 x 14 mm |
Each for $157.10
|
|
|||||
19-174-805
|
Ampco Safety Tools
WO13X15 |
13 x 15 mm |
Each for $158.10
|
|
|||||
19-174-806
|
Ampco Safety Tools
WO14X15 |
14 x 15 mm |
Each for $180.60
|
|
|||||
19-174-807
|
Ampco Safety Tools
WO14X16 |
14 x 16 mm |
Each for $180.60
|
|
|||||
19-174-808
|
Ampco Safety Tools
WO14X17 |
14 x 17 mm |
Each for $180.60
|
|
|||||
19-174-812
|
Ampco Safety Tools
WO16X17 |
16 x 17 mm |
Each for $183.60
|
|
|||||
19-174-813
|
Ampco Safety Tools
WO16X18 |
16 x 18 mm |
Each for $185.70
|
|
|||||
19-174-816
|
Ampco Safety Tools
WO17X19 |
17 x 19 mm |
Each for $186.70
|
|
|||||
19-174-817
|
Ampco Safety Tools
WO17X22 |
17 x 22 mm |
Each for $189.80
|
|
|||||
19-174-818
|
Ampco Safety Tools
WO18X19 |
18 x 19 mm |
Each for $186.70
|
|
|||||
19-174-819
|
Ampco Safety Tools
WO18X20 |
18 x 20 mm |
Each for $186.70
|
|
|||||
19-174-820
|
Ampco Safety Tools
WO18X22 |
18 x 22 mm |
Each for $186.70
|
|
|||||
19-174-821
|
Ampco Safety Tools
WO19X22 |
19 x 22 mm |
Each for $189.80
|
|
|||||
19-174-822
|
Ampco Safety Tools
WO19X24 |
19 x 24 mm |
Each for $189.80
|
|
|||||
19-174-824
|
Ampco Safety Tools
WO20X22 |
20 x 22 mm |
Each for $189.80
|
|
|||||
19-174-828
|
Ampco Safety Tools
WO21X22 |
21 x 22 mm |
Each for $194.90
|
|
|||||
19-174-829
|
Ampco Safety Tools
WO21X23 |
21 x 23 mm |
Each for $194.90
|
|
|||||
19-174-830
|
Ampco Safety Tools
WO22X23 |
22 x 23 mm |
Each for $194.90
|
|
|||||
19-174-831
|
Ampco Safety Tools
WO22X24 |
22 x 24 mm |
Each for $194.90
|
|
|||||
19-174-832
|
Ampco Safety Tools
WO22X27 |
22 x 27 mm |
Each for $215.50
|
|
|||||
19-174-834
|
Ampco Safety Tools
WO23X26 |
23 x 26 mm |
Each for $213.50
|
|
|||||
19-174-835
|
Ampco Safety Tools
WO24X26 |
24 x 26 mm |
Each for $215.50
|
|
|||||
19-174-836
|
Ampco Safety Tools
WO24X27 |
24 x 27 mm |
Each for $215.50
|
|
|||||
19-174-837
|
Ampco Safety Tools
WO24X30 |
24 x 30 mm |
Each for $266.50
|
|
|||||
19-174-840
|
Ampco Safety Tools
WO25X28 |
25 x 28 mm |
Each for $266.50
|
|
|||||
19-174-841
|
Ampco Safety Tools
WO26X29 |
26 x 29 mm |
Each for $278.50
|
|
|||||
19-174-842
|
Ampco Safety Tools
WO27X30 |
27 x 30 mm |
Each for $280.50
|
|
|||||
19-174-843
|
Ampco Safety Tools
WO27X32 |
27 x 32 mm |
Each for $282.00
|
|
|||||
19-174-846
|
Ampco Safety Tools
WO30X32 |
30 x 32 mm |
Each for $282.00
|
|
|||||
19-174-847
|
Ampco Safety Tools
WO30X34 |
30 x 34 mm |
Each for $295.00
|
|
|||||
19-174-848
|
Ampco Safety Tools
WO30X36 |
30 x 36 mm |
Each for $314.50
|
|
|||||
19-174-849
|
Ampco Safety Tools
WO32X36 |
32 x 36 mm |
Each for $316.50
|
|
|||||
19-174-850
|
Ampco Safety Tools
WO34X36 |
34 x 36 mm |
Each for $316.50
|
|
|||||
19-174-851
|
Ampco Safety Tools
WO36X41 |
36 x 41 mm |
Each for $481.50
|
|
|||||
19-174-852
|
Ampco Safety Tools
WO41X46 |
41 x 46 mm |
Each for $505.00
|
|
|||||
19-174-853
|
Ampco Safety Tools
WO46X50 |
46 x 50 mm |
Each for $574.50
|
|
|||||
19-174-857
|
Ampco Safety Tools
WO50X55 |
50 x 55 mm |
Each for $612.00
|
|
|||||
19-174-858
|
Ampco Safety Tools
WO50X60 |
50 x 60 mm |
Each for $1,357.00
|
|
|||||
19-174-859
|
Ampco Safety Tools
WO55X60 |
55 x 60 mm |
Each for $1,546.00
|
|
|||||
19-174-860
|
Ampco Safety Tools
WO65X70 |
65 x 70 mm |
Each for $1,718.00
|
|
|||||
19-174-861
|
Ampco Safety Tools
WO6X7 |
6 x 7 mm |
Each for $80.60
|
|
|||||
19-174-862
|
Ampco Safety Tools
WO6X8 |
6 x 8 mm |
Each for $82.70
|
|
|||||
19-174-866
|
Ampco Safety Tools
WO70X75 |
70 x 75 mm |
Each for $1,718.00
|
|
|||||
19-174-867
|
Ampco Safety Tools
WO7X8 |
7 x 8 mm |
Each for $86.70
|
|
|||||
19-174-868
|
Ampco Safety Tools
WO7X9 |
7 x 9 mm |
Each for $123.50
|
|
|||||
19-174-869
|
Ampco Safety Tools
WO8X10 |
8 x 10 mm |
Each for $123.50
|
|
|||||
19-174-870
|
Ampco Safety Tools
WO8X9 |
8 x 9 mm |
Each for $123.50
|
|
|||||
19-174-873
|
Ampco Safety Tools
WO9X10 |
9 x 10 mm |
Each for $123.50
|
|
|||||
19-174-874
|
Ampco Safety Tools
WO9X11 |
9 x 11 mm |
Each for $123.50
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Double Open Wrench | |
Aluminum Bronze | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.