Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caresfield Direct Thermal Label Roll, 1 in. Core
Suitable for applications not requiring a long shelf life
Supplier: Caresfield 82077
This item is not returnable.
View return policy
Description
- No ink, toner or ribbon required
Specifications
Permanent, Pressure-sensitive | |
Matte White | |
Labels | |
2 in. |
Black on White | |
Label | |
Non-sterile | |
1 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction