Learn More
Andwin Scientific COTTON PADS 20 PK/CS
Supplier: Andwin Scientific 21902985CS
Cotton Pads Useful for the printing industry. All-purpose 100% cotton. No binders or chemicals. Excellent wet strength. Will not scratch the most delicate coatings or surfaces. Will not contaminate developing solutions or chemicals. 10x10cm