Learn More
World Precision Instrument Room Temperature Vulcanizing Adhesive
Supplier: World Precision Instrument 1571
Clear silicone sealant with good bonding to plastic
This room temperature vulcanizing (RTV) adhesive has an acyloxy/moisture curing system that bonds well with many surfaces. After curing, it will withstand -55 to 200°C. No mixing is required. This is a handy, general purpose laboratory sealant. (Acetic acid is a by-product during curing.)
This item is not returnable.
View return policy