Learn More
Chemglass Life Sciences Cap, Chem Snap, 11mm, Clear, Starburst Septum
SDP
Supplier:  Chemglass Life Sciences CV35010011
Encompass_Preferred
Cap, Chem Snap, 11mm, Clear, Starburst Septum. Chem Snap Caps may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. The starburst allows for easier needle penetration. These caps were developed to handle the most volatile sample providing a liquid airtight seal to the vial. Simply snap on the cap and an evaporation-proof seal of a crimp cap is achieved. Snap caps are color coded for easy sample identification.
Catalog No. 50-153-2554