Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Vinegar Eel
Lives in vinegar on the fungus.
Supplier: Carolina Biological Supply 133266
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
They can be observed under the microscope at 20X.
- Turbatrix aceti
- Shipped in a 1 oz. liquid culture
- Enough materials for 30 students
Specifications
Ea. | |
Turbatrix aceti | |
Class of 30 students | |
Vinegar Eel |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction