Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, pH 12.45, MilliporeSigma™
Supplier: MilliporeSigma EM01802
Specifications
500 mL | |
Plastic Bottle |
Safety and Handling
Recommended Storage : Store at 15 to 30°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction