Learn More
Chemglass Life Sciences Bonded Closure, Standard, 9mm, Green, Open Hole, 0.40" PTFE/Silicone with Slit
Supplier: Chemglass Life Sciences CV3845BSG9
Bonded Closure, Standard, 9mm, Green, Open Hole, 0.40" PTFE/Silicone with Slit. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.