Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Click to view available options
Holds:
28 x 10 mm dia. Block
46 x 6 mm dia. block
Description
- Aluminum alloy blocks
Specifications
For Use With (Equipment) | Digital and Touch Screen Dry Baths/Block Heaters (No compact models) |
Holds | 28 x 10 mm dia. Block |
No. of Places | 28 |
Type | 28 x 10 mm dia. Block |
Diameter (Metric) | 10 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction