Learn More
Agilent Technologies BL21CODONPLUS(DE3)RILX CELLS
Supplier:  Agilent Technologies 626616637_818464089
Encompass
BL21-CodonPlus(DE3)-RIL-X Competent Cells encodes T7 RNA polymerase under control of the lacUV5 promoter for easy protein expression plus methionine label. Use with pET or pCAL vectors. Used for labeling cells of supercoiled DNA.
Catalog No. 50-125-354
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.