Learn More
PBL Assay Science Anti-Mouse Interferon Alpha (IFN-Alpha) Rat Monoclonal Antibody, Clone RMMA-1 (MAb) [250 micrograms (ug)]
Supplier: PBL Assay Science 221001
Rat monoclonal antibody Anti-Mouse Interferon Alpha, Clone RMMA-1 is manufactured by PBL Assay Science (Catalog #22100-1) for research use only (RUO) and packaged in 250 micrograms (ug). This MAb binds to and neutralizes mouse IFN alpha. It is suitable for use in neutralization and ELISA applications.