Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD8, BUV395; Clone: RPA-T8; BD
$224.00 - $456.00
Specifications
Antigen | CD8 |
---|---|
Clone | RPA-T8 |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 395 |
Host Species | Mouse |
Description
Host Species: Mouse
Clone: RPA-T8
Isotype: IgG1, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Flow cytometry
Specifications
CD8 | |
Monoclonal | |
Mouse | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
Affinity Purified |
RPA-T8 | |
Brilliant Ultraviolet 395 | |
RUO | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title