Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-CA72-4, 0.2mg
Supplier: Advanced ImmunoChemical, Inc. 2CA720.2MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Tumor Markers / Monoclonal mouse anti-CA72-4. MAbs: 7C1,1C2,2C3.Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native CA72-4. MAb isotypes: IgG1 for MAbs 7C1,1C2,2C3Applications:Detection of CA72-4 antigen. Recommended pairs for sandwich immunoassay (capture – detection):1C2 – 7C12C3 – 7C1All MAbs are working in Western blotting. Purifications: Ion exchange chromatographyPresentationPBS, pH 7.4, 0.09 % sodium azide (NaN₃)Storage Conditions:+4 °C (+2 … +8 °C allowed)Material Safety Note:This product is sold for research use only. Standard Laboratory Practices should be followed when handling this material.Product contains sodium azide as a preservative. Although the amount of sodium azide is very small appropriate care must be taken when handling this product.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.