Learn More
United Scientific™ All Glass Vacuum Filter Holder Set and Accessories
All Glass Vacuum Filter Holder Set and Accessories used in particulate contamination analysis.
Supplier: United Scientific™ FHGF300
Description
- Used in particulate contamination analysis for filtering aqueous, organic, or corrosive liquids.
- Recommended for HPLC solvent filtration.
- Complete set includes a borosilicate glass funnel, anodized aluminum spring clamp, vacuum base with sintered disc, and ground joint flask.
- Membrane filters are not included.
- Parts are also available separately.
Specifications
Particulate Contamination Analysis | |
Borosilicate Glass | |
34/35 |
Glass Funnel | |
Replacement Funnel, 300 mL | |
300 mL |