Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-PACK™ Unit Dose Test Cups
Specially prepared reagents for a variety of assays using Tosoh AIA systems
$1327.00
Catalog Number | Mfr. No. | Detectable Analytes | For Use With (Application) | Range | Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | For Use With (Application) | Range | Type | Price | Quantity & Availability | ||
21-280-022
|
Tosoh Bioscience
025287 |
Cortisol | Measurement of Serum Cortisol and evaluate hypothalamic-pituitaryadrenal function as well as clinical diagnosis | Up to 60μg/dL | Cortisol Assay Test Cup |
Each for $1,327.00
|
|
21-280-024
|
Tosoh Bioscience
025266 |
Human Growth Hormone | Screening Tests for HGH deficiency, HGH hypersecretion and assessment of pituitary-hypothalamic function | Up to 50ng/mL | HGH Assay Test Cup |
Each for $1,409.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
21-280-025
|
Tosoh Bioscience
025260 |
Insulin | Fundamental defects in Human diabetes | Up to 320mU/mL | Insulin Assay Test Cup |
Each for $1,409.00
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
21-280-021
|
Tosoh Bioscience
025259 |
beta-2-microglobulin | Serum BMG determination | Up to 0.4mg/L | BMG (beta-2-Microglobulin) Assay Test Cup |
Each for $1,409.00
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
21-280-023
|
Tosoh Bioscience
025284 |
C-Peptide | C-Peptide measurements are also used as an additional means of evaluating Glucose tolerance Tests | Up to 30ng/mL serum, 300ng/mL urine | C-Peptide Assay Test Cup | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
For determination of analytes using Tosoh Automated Immunoassay Systems.
- Contain all necessary components for analysis
- Single unit dose requires no preprocessing
- Format: Dry reagent with 90 day calibration stability for most assays
- Designed for 10 minute incubation protocol
- Shelf life: One year from date of manufacture
- Bar-coded for easy identification and inventory
Order Info
Packs of 100 (20 cups x 5 trays).
Specifications
AIA Test Cup | |
100 Pack |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title