Learn More
Across International Ai 200°C 7L SST Compact Desktop Heated Recirculator 220V
SDP
Supplier:  Across International H2007SST.220
Encompass_Preferred
The new Ai H200 stainless steel series heated recirculators with 2,800 watts heater offer outstanding performance and high reliability as well as quiet operation and ease-of-use. Powerful force/suction pump provides constant flow rates, while the 2nd generation PID controller gives you the ideal solution when you need precise temperature control.
Catalog No. 50-225-1353