Learn More
Metrohm USA Petri Dishes, 100 x 15 mm, Sterile, 500/Pk
Supplier: Metrohm USA GFSIC5785
a polypropylene microcentrifuge tube rack that holds 0.5mL tubes. It has a 96-well format with alphanumeric coordinates to identify each well location. The rack is made from durable autoclavable polypropylene and can withstand temperatures from -20°C to 121°C. It has an external height of 38mm, length of 127mm, width of 85mm, and well depth of 28mm. This microtube rack allows for organized storage and easy retrieval of samples in 0.5mL tubes. Its sturdy construction and alphanumeric grid ensure sample tracking and identification.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.