Learn More
3M™ GVP-Series PAPR Accessory, Flowmeter
For use with GVP-Series PAPRs
Supplier: 3M™ 7000126094
Description
- These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
- 3M Catalog# GVP-113
Specifications
Reusable | |
Measures The System Airflow to Make Certain That it is at The Proper Level for The Headgear and Cartridge Being Used | |
GVP-113 |
Flowmeter | |
GVP-Series Powered Air Purifying Respirator | |
1 Ea. |