Learn More
2-Mercapto-5-methoxybenzimidazole, 99%, Thermo Scientific Chemicals
Description
2-Mercapto-5-methoxybenzimidazole is used in the preparation of COX-2 inhibitors. It is also employed in the preparation of compounds such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole. Further, it acts as a hetercocyclic building block and serves as an intermediate of drug omeprazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Mercapto-5-methoxybenzimidazole is used in the preparation of COX-2 inhibitors. It is also employed in the preparation of compounds such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole. Further, it acts as a hetercocyclic building block and serves as an intermediate of drug omeprazole.
Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in methanol. Insoluble in water.
Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
Specifications
Melting Point | 255°C to 259°C |
Quantity | 100 g |
Beilstein | 3605123 |
Solubility Information | Soluble in dimethyl sulfoxide. Slightly soluble in methanol. Insoluble in water. |
IUPAC Name | 5-methoxy-1,3-dihydrobenzimidazole-2-thione |
Formula Weight | 180.23 |
Percent Purity | 99% |
Chemical Name or Material | 2-Mercapto-5-methoxybenzimidazole |
Safety and Handling
Hazard Category | H302-H317 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c |
EINECSNumber | 253-326-2 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.