Assay Details
Target Gene Details
Entrez Gene ID: | 268566 |
Gene Name: | gephyrin |
Gene Aliases: |
5730552E08Rik, AI662856, BC027112, C230040D23, GPH, GPHRYN, geph |
Location: |
Chr.12:78224153-78684772 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gphn | NM_145965.2 | 17 | 1822 | NP_666077.2 |
NM_172952.3 | 18 | 1813 | NP_766540.2 | |
AK081716.1 | 1 | 2802 | ||
AK082353.1 | 18 | 1735 | BAC38476.1 | |
AK144569.1 | 17 | 1824 | ||
BC127058.1 | 17 | 1744 |