Assay Details
Target Gene Details
Entrez Gene ID: | 18191 |
Gene Name: | neurexin III |
Gene Aliases: |
- |
Location: |
Chr.12:88722865-90334935 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrxn3 | NM_001198587.3 | 17 | 4340 | NP_001185516.2 |
NM_001252074.2 | 4 | 1555 | NP_001239003.1 | |
NM_172544.3 | 17 | 3257 | NP_766132.2 | |
AK032126.1 | 17 | 3249 | BAC27716.1 | |
AK042730.1 | 4 | 1557 | BAC31344.1 | |
AK129205.1 | BAC98015.2 | |||
BC060719.1 | 18 | 3915 | AAH60719.1 |