Learn More
ZymeTx ZstatFlu™ Test for Influenza Types A and B
Test is simple and easy to perform
Specifications
Detectable Analytes Influenza A and B
View More Specs
Description
Description
  • Test results within 20 minutes
  • CLIA Complexity: Waived
  • An endogenous viral-encoded enzyme assay for the qualitative determination of influenza Types A and B from throat swab specimens
  • Test requires no special handling or reading instruments
  • Simply obtain specimen, combine with diluent, incubate, add stop reagent and read results
Specifications
Specifications
Influenza A and B