Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Nitrocellulose Membrane Filters
Recommended for the majority of routine applications, manufactured under strictly controlled conditions
Supplier: Cytiva 7195009
Description
- Filters have a noticeably improved flexibility
- Made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity
- Membranes are among the strongest of their type available, as measured and compared by burst pressure tests
- The residue after filtration is found to be almost completely on the surface of the membrane and allows physical recovery of deposits and microscopic examination
- Low level of extractables, generally below that of other membranes of a similar type
- The narrow pore size distribution is closely controlled due to the advanced manufacturing and control system
- Batch-to-batch variation is minimized providing more consistent laboratory results
- Autoclave at 121°C without any shrinkage or loss of integrity
Specifications
Autoclavable | |
Nitrocellulose | |
5 μm | |
Sample Preparation, Microbiological Studies, and Filtration of Aqueous Solutions | |
Membrane Filter |
White | |
Nitrocellulose | |
Non-sterile | |
25/Pk. | |
90 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction