Learn More
US Biomax Inc TMABO2081 BONE DISEASE SPECTRM
SDP
Supplier:  US Biomax Inc BO2081
Encompass
BO2081 Bone disease spectrum (Osteosarcoma,Chondrosarcoma,Myeloma,Ewing's sarcoma,Chordoma,Adamantinoma of long bones,Adamantinoma of mandible,Osteoblastoma,Giant cell tumor,Chondroma,Fibrous dysplasia,Aneurysmal bone cyst) ) and cancer ajdacent normal bone tissue array, 104 cases/208 cores, unstained slide
Catalog No. NC2137412