Learn More
Ctech Glass TLC SPOTTING CAPILLARY TUBES
SDP
Supplier:  Ctech Glass CT9501
Encompass
Glass capillary tubes ideal for spotting TLC chromatographic applications and melting point determination experiments. Low grade borosilicate glass. Stand up to 110 C degrees. ID: 0.3mm, OD: 0.54mm, variation: 0.01~0.015mm. Length: 100mm. Two ends opened. 1000 pcs/pkg (aprox).
Catalog No. NC0962445
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.