Learn More
Sartorius Microsart™ e.motion Membrane Filters
Membrane Filters suitable for use in the Microsart e.motion dispenser
$260.00 - $339.00
Catalog Number | Mfr. No. | Color | Diameter (Metric) | Pore Size | Grid Color | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Diameter (Metric) | Pore Size | Grid Color | Price | Quantity & Availability | ||
5090117778
|
Sartorius
13806Z50SCM |
Green | 50 mm | 0.45 μm | Dark Green |
Pack of 300 for $260.00
|
|
||
5090117755
|
Sartorius
13004Z47SCM |
Gray | 47 mm | 0.8 μm | White |
Pack of 300 for $260.00
|
|
||
5090117784
|
Sartorius
13906Z50SCM |
White | 50 mm | 0.45 μm | Green |
Pack of 300 for $260.00
|
|
||
5090117765
|
Sartorius
13006Z50SCM |
Gray | 50 mm | 0.45 μm | White |
Pack of 300 for $260.00
|
|
||
14-555-580
|
Sartorius
11402Z47SCM |
White | 47 mm | 0.3 μm | Black |
Pack of 300 for $260.00
|
|
||
14-555-581
|
Sartorius
11402Z47SFM |
White | 47 mm | 3.0 μm | Black |
Pack of 300 for $339.00
|
|
||
14-555-605
|
Sartorius
11406Z50SCM |
White | 50 mm | 0.45 μm | Black |
Pack of 300 for $260.00
|
|
||
14-555-604
|
Sartorius
11406Z47SCM |
White | 47 mm | 0.45 μm | Black |
Pack of 300 for $332.00
|
|
||
14-555-610
|
Sartorius
11407Z47SCM |
White | 47 mm | 0.2 μm | Black |
Pack of 300 for $260.00
|
|
||
14-555-628
|
Sartorius
13006Z47SCM |
Gray | 47 mm | 0.45 μm | White |
Pack of 300 for $260.00
|
|
||
14-555-650
|
Sartorius
13806Z47SCM |
Dark Green | 47 mm | 0.45 μm | Green |
Pack of 300 for $260.00
|
|
||
14-555-656
|
Sartorius
13906Z47SCM |
White | 47 mm | 0.45 μm | Green |
Pack of 300 for $260.00
|
|
||
14-555-619
|
Sartorius
13004Z50SCM |
Gray | 50 mm | 0.8 μm | White | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | ||
Description
Membrane Filters suitable for use in the Microsart e.motion dispenser
The membrane filters suitable for use in the Microsart™ e.motion dispenser are sterile-sealed, without protective paper on top of each filter, in a specially designed individual package on a band. The special pleating of the band of membrane filter units ensures that they are perfectly flat when dispensed.The shape of the sealed band guarantees uniform dispensing of the individual membrane filters.The Microsart™ e.motion Membrane Filters are also ideal suitable for all users of Sartorius Nutrient Pad Sets: Most of the NPS types are available with membranes for dispensers.
- outstanding recovery rates for microorganisms
- 0.45μm are acc. to ISO 7704
- Multi-fit: Fits into other dispensers
- protective paper-free
- packaged on a special pleated band
- product data are printed on
- High Flow membranes available
- gamma irradiated, 25kGray
47 or 50mm in diameter, white, grey or green and gridded
Growth Promotion Test acc. to ISO 7704
Specifications
Cellulose Nitrate | |
Sterile |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.