Learn More
AUTOMATION TRAINER P50 Beckman-Type AP96, Clear, Filtered, Sterile, Racked
SDP
Supplier:  AUTOMATION TRAINER BP50CLFSR
Encompass_Preferred
This item has a minimum order quantity of 10 per supplier requirements.
Automation Trainer's pipette tips are designed to be a direct replacement to Beckman's AP96 P20, P50, & P250 tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required.- Fits Biomek 3000, NX, NXp, FX and FXp Laboratory Automation Workstations- High-Grade Polypropylene Build- P20 (Case of 50 96-Tip Racks), P50 (Case of 50 96-Tip Racks), and P250 (Case of 50 96-Tip Racks)
Catalog No. NC2219000