Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NVP ADW 742
ATP-competitive inhibitor of IGF1R
$280.72 - $1173.92
Specifications
Purity | >99% |
---|---|
Product Type | NVP ADW 742 |
Molecular Weight (g/mol) | 453.58 |
Inhibitors | NVP ADW 742 |
Description
ATP-competitive inhibitor of IGF1R (IC50 in the range of 0.1-0.2 μM). Also inhibits c-Kit kinase activity (IC50 in the range of 3-5 μM). Exhibits a synergistic effect with Imatinib, enhancing the sensitivity of multiple small cell lung cancer cell lines to the chemotherapeutics etoposide (Cat. No. 1226) and carboplatin (Cat. No. 2626).Specifications
>99% | |
NVP ADW 742 | |
453.58 | |
NVP ADW 742 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title