Learn More
NQO1 Mouse anti-Human, CoraLite 488, Clone: 1E5G7, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL48867240100UL
This item is not returnable.
View return policy
Description
The NQO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NQO1. This antibody recognizes human, rat antigen. The NQO1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
NQO1 | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
1728 | |
IgG2a |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
1E5G7 | |
Mouse | |
Primary | |
Human, Rat |